ENA152121111 Enamine

[2-(4-fluorophenyl)-1,3-oxazol-4-yl]acetonitrile

  • MDL number MFCD09863268
  • Cas Number : 22087-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28991

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28991
Availability details for
Quantity
  • Availability
  • Add to Cart