ENA152121114 Enamine

4,5-dimethyl-2-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD08443345
  • Cas Number : 838840-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29200
Availability details for
Quantity
  • Availability
  • Add to Cart