ENA152121115 Enamine

methyl 1,2,5-trimethyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD00203867
  • Cas Number : 14186-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30443

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-30443
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart