ENA152121118 Enamine

1-[4-(difluoromethoxy)phenyl]ethan-1-amine

  • MDL number MFCD09028381
  • Cas Number : 136123-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30453

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-30453
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart