ENA152121124 Enamine

1-[5-(4-fluorophenyl)furan-2-yl]methanamine

  • MDL number MFCD08444637
  • Cas Number : 923230-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29404
Availability details for
Quantity
  • Availability
  • Add to Cart