ENA152121133 Enamine

N-cyclohexylpiperazine-1-carboxamide hydrochloride

  • MDL number MFCD09863347
  • Cas Number : 1094600-70-0
  • Molecular Weight : 247.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30474
Availability details for
Quantity
  • Availability
  • Add to Cart