ENA152121136 Enamine

3-methylquinoxaline-2-thiol

  • MDL number MFCD00546511
  • Cas Number : 58773-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03198

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03198
Availability details for
Quantity
  • Availability
  • Add to Cart