ENA152121142 Enamine

2-(4-fluorophenyl)morpholine

  • MDL number MFCD09863353
  • Cas Number : 62243-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-60
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30487

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-30487
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart