ENA152121144 Enamine

2-(morpholin-4-yl)-2-phenylethan-1-amine

  • MDL number MFCD01631944
  • Cas Number : 31466-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30489
Availability details for
Quantity
  • Availability
  • Add to Cart