ENA152121145 Enamine

2,6-dimethyl-3-nitroaniline

  • MDL number MFCD00156348
  • Cas Number : 67083-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80...82
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30490

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-30490
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart