ENA152121146 Enamine

2-(morpholin-4-ylsulfonyl)aniline

  • MDL number MFCD04035364
  • Cas Number : 208643-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30491

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30491
Availability details for
Quantity
  • Availability
  • Add to Cart