ENA152121149 Enamine

N-cyclohexyl-N-methylsulfamoyl chloride

  • MDL number MFCD08443401
  • Cas Number : 83842-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30496
Availability details for
Quantity
  • Availability
  • Add to Cart