ENA152121163 Enamine

N-(4-amino-2,6-dichlorophenyl)-4-methanesulfonylbenzamide

  • MDL number MFCD09045324
  • Cas Number : 926207-01-4
  • Molecular Weight : 359.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-29629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29629
Availability details for
Quantity
  • Availability
  • Add to Cart