ENA152121165 Enamine

2-(4-nitro-1H-imidazol-1-yl)acetamide

  • MDL number MFCD00621267
  • Cas Number : 120399-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30520
Availability details for
Quantity
  • Availability
  • Add to Cart