ENA152121170 Enamine

N-[2-(morpholin-4-yl)ethyl]piperidine-4-carboxamide

  • MDL number MFCD03137409
  • Cas Number : 429637-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30534
Availability details for
Quantity
  • Availability
  • Add to Cart