ENA152121173 Enamine

2,3,4,9-tetrahydro-1H-carbazole-8-carboxylic acid

  • MDL number MFCD00458334
  • Cas Number : 65764-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30538
Availability details for
Quantity
  • Availability
  • Add to Cart