ENA152121181 Enamine

4-methoxy-3-[(pyridin-4-yl)methoxy]benzoic acid

  • MDL number MFCD08442453
  • Cas Number : 187970-70-3
  • Molecular Weight : 259.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30547
Availability details for
Quantity
  • Availability
  • Add to Cart