ENA152121189 Enamine

methyl 3-(chlorosulfonyl)-4-methylbenzoate

  • MDL number MFCD09746474
  • Cas Number : 372198-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30560

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-30560
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart