ENA152121190 Enamine

2-bromo-4-methylbenzenesulfonyl chloride

  • MDL number MFCD08445068
  • Cas Number : 89794-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30562

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30562
Availability details for
Quantity
  • Availability
  • Add to Cart