ENA152121194 Enamine

[3-chloro-4-(difluoromethoxy)-5-methoxyphenyl]methanol

  • MDL number MFCD08442639
  • Cas Number : 926183-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29714
Availability details for
Quantity
  • Availability
  • Add to Cart