ENA152121198 Enamine

(6-methoxynaphthalen-2-yl)methanol

  • MDL number MFCD06202657
  • Cas Number : 60201-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29715
Availability details for
Quantity
  • Availability
  • Add to Cart