ENA152121207 Enamine

tetrahydrothiophene-3-carbonitrile 1,1-dioxide

  • MDL number MFCD06809749
  • Cas Number : 17389-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30588

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30588
Availability details for
Quantity
  • Availability
  • Add to Cart