ENA152121209 Enamine

pteridin-4(3H)-one

  • MDL number MFCD00039725
  • Cas Number : 700-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30591

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30591
Availability details for
Quantity
  • Availability
  • Add to Cart