ENA152121210 Enamine

5-[(2-amino-1,3-thiazol-4-yl)methyl]-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione

  • MDL number MFCD05719131
  • Cas Number : 690642-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30592

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30592
Availability details for
Quantity
  • Availability
  • Add to Cart