ENA152121234 Enamine

2-amino-N-ethylbenzenesulfonamide

  • MDL number MFCD09732459
  • Cas Number : 98489-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30623

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30623
Availability details for
Quantity
  • Availability
  • Add to Cart