ENA152121242 Enamine

4-(cyclopropylsulfamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD08899153
  • Cas Number : 1000932-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30638
Availability details for
Quantity
  • Availability
  • Add to Cart