ENA152121247 Enamine

5-(pyrrolidine-1-carbonyl)-1H-pyrrole-3-sulfonyl chloride

  • MDL number MFCD09863284
  • Cas Number : 950094-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29737
Availability details for
Quantity
  • Availability
  • Add to Cart