ENA152121250 Enamine

{[1-(difluoromethyl)-1H-imidazol-2-yl]methyl}(methyl)amine

  • MDL number MFCD09040727
  • Cas Number : 929972-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30649
Availability details for
Quantity
  • Availability
  • Add to Cart