ENA152121271 Enamine

2-cyclopentyl-1-(piperazin-1-yl)ethan-1-one

  • MDL number MFCD08442402
  • Cas Number : 926238-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30681
Availability details for
Quantity
  • Availability
  • Add to Cart