ENA152121280 Enamine

3-[(pyrimidin-2-yl)amino]benzoic acid

  • MDL number MFCD08445169
  • Cas Number : 198195-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30692
Availability details for
Quantity
  • Availability
  • Add to Cart