ENA152121282 Enamine

(pyrimidin-2-ylamino)acetic acid

  • MDL number MFCD03007289
  • Cas Number : 79858-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30699

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-30699
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart