ENA152121286 Enamine

N-(cyclopropylmethyl)-N-propylsulfamoyl chloride

  • MDL number MFCD08443299
  • Cas Number : 926207-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30703
Availability details for
Quantity
  • Availability
  • Add to Cart