ENA152121291 Enamine

bis(2-methoxyethyl)sulfamoyl chloride

  • MDL number MFCD08445094
  • Cas Number : 371150-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30708

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30708
Availability details for
Quantity
  • Availability
  • Add to Cart