ENA152121292 Enamine

1-(2,2-dimethylpropanoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00179481
  • Cas Number : 114525-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136...138
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30709

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30709
Availability details for
Quantity
  • Availability
  • Add to Cart