ENA152121303 Enamine

3-[(chlorosulfonyl)(methyl)amino]propanenitrile

  • MDL number MFCD08443011
  • Cas Number : 209971-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30732
Availability details for
Quantity
  • Availability
  • Add to Cart