ENA152121312 Enamine

4-amino-N-(4-hydroxycyclohexyl)benzene-1-sulfonamide

  • MDL number MFCD09733638
  • Cas Number : 174628-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30746
Availability details for
Quantity
  • Availability
  • Add to Cart