ENA152121313 Enamine

3,4-dichloro-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD00617272
  • Cas Number : 34576-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 278-280
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30747
Availability details for
Quantity
  • Availability
  • Add to Cart