ENA152121318 Enamine

1-benzyl-3-(2-methylphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD08443505
  • Cas Number : 926246-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29775
Availability details for
Quantity
  • Availability
  • Add to Cart