ENA152121319 Enamine

methyl 4-fluoro-1H-indole-2-carboxylate

  • MDL number MFCD06653401
  • Cas Number : 113162-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30755
Availability details for
Quantity
  • Availability
  • Add to Cart