ENA152121333 Enamine

1-[1-(2-chlorophenyl)ethyl]piperazine

  • MDL number MFCD05186117
  • Cas Number : 512775-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30772
Availability details for
Quantity
  • Availability
  • Add to Cart