ENA152121336 Enamine

2-(4-methyl-1,3-thiazol-2-yl)ethan-1-amine

  • MDL number MFCD05221349
  • Cas Number : 61887-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30778
Availability details for
Quantity
  • Availability
  • Add to Cart