ENA152121343 Enamine

2-(1,4-diazepan-1-yl)-1-(pyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD09045356
  • Cas Number : 55829-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30790
Availability details for
Quantity
  • Availability
  • Add to Cart