ENA152121345 Enamine

N-(4-amino-2-chlorophenyl)methanesulfonamide

  • MDL number MFCD09048008
  • Cas Number : 18735-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30798
Availability details for
Quantity
  • Availability
  • Add to Cart