ENA152121346 Enamine

5-amino-2-chlorobenzamide

  • MDL number MFCD07787477
  • Cas Number : 111362-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153...155
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30799

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-30799
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart