ENA152121353 Enamine

1-[3-(1H-tetrazol-1-yl)phenyl]ethanol

  • MDL number MFCD08443017
  • Cas Number : 926189-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29795
Availability details for
Quantity
  • Availability
  • Add to Cart