ENA152121354 Enamine

6-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}pyridin-3-amine

  • MDL number MFCD09050249
  • Cas Number : 926211-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29796
Availability details for
Quantity
  • Availability
  • Add to Cart