ENA155457778 Enamine

4-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09863423
  • Cas Number : 1000932-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-30825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30825
Availability details for
Quantity
  • Availability
  • Add to Cart