ENA155457782 Enamine

2-(chloromethyl)-3-formylindolizine-1-carbonitrile

  • MDL number MFCD09863433
  • Cas Number : 1000932-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30860
Availability details for
Quantity
  • Availability
  • Add to Cart