ENA155457783 Enamine

N-(pyridin-2-ylmethyl)sulfamide

  • MDL number MFCD09863434
  • Cas Number : 924307-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30863
Availability details for
Quantity
  • Availability
  • Add to Cart