ENA155457784 Enamine

1-chloro-3-(chloromethyl)pyrido[1,2-a]benzimidazole-4-carbonitrile

  • MDL number MFCD09863436
  • Cas Number : 1000932-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30871
Availability details for
Quantity
  • Availability
  • Add to Cart